Product Details

SNP ID
rs199659066
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:172445189 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTATTAATACTAGATTCTGTCCAG[A/G]CCCGAGAACTTTCATCTTTCAGAGT
Phenotype
MIM: 601898
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
GHSR PubMed Links

Gene Details

Gene
GHSR
Gene Name
growth hormone secretagogue receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004122.2 1116 Intron NP_004113.1
NM_198407.2 1116 Missense Mutation GCC,GTC A358V NP_940799.1

View Full Product Details