Product Details

SNP ID
rs200913424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:68930043 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGACTGCTCGATCTAGGGGCTTTA[C/T]AGGTTGATCATGGTGAATTCTTGAT
Phenotype
MIM: 616382
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UGT2A3 PubMed Links

Gene Details

Gene
UGT2A3
Gene Name
UDP glucuronosyltransferase family 2 member A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024743.3 1385 Missense Mutation ATA,GTA I452V NP_079019.3
XM_011532247.2 1385 Missense Mutation ATA,GTA I458V XP_011530549.1

View Full Product Details