Product Details
- SNP ID
-
rs200219137
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:10443106 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTTTAAAGAGTAACTGTTTACTT[A/G]TTTGCCCTTGGAGGAAAGAACATCC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF518B
PubMed Links
Gene Details
- Gene
- ZNF518B
- Gene Name
- zinc finger protein 518B
There are no transcripts associated with this gene.
View Full Product Details