Product Details

SNP ID
rs200257636
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:160940 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTTCTCATTTCACCCAAAACCT[G/T]TGGACAGTGCAGGGCATAGAAGATT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF718 PubMed Links

Gene Details

Gene
ZNF718
Gene Name
zinc finger protein 718
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039127.5 1073 Silent Mutation CTG,CTT L85L NP_001034216.2
NM_001289930.1 1073 Silent Mutation CTG,CTT L53L NP_001276859.1
NM_001289931.1 1073 Intron NP_001276860.1
XM_017007979.1 1073 Intron XP_016863468.1
XM_017007980.1 1073 Silent Mutation CTG,CTT L53L XP_016863469.1
XM_017007981.1 1073 Silent Mutation CTG,CTT L26L XP_016863470.1

View Full Product Details