Product Details
- SNP ID
-
rs201929602
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:88731387 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGAAGTTTCTTTCGAATCCTTTTC[C/T]TTTCTTCTCTCATTTCCTGGAGGTG
- Phenotype
-
MIM: 613299
MIM: 613300
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FAM13A
PubMed Links
Gene Details
- Gene
- FAM13A
- Gene Name
- family with sequence similarity 13 member A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001015045.2 |
4805 |
Missense Mutation |
AAG,AGG |
K636R |
NP_001015045.1 |
NM_001265578.1 |
4805 |
Missense Mutation |
AAG,AGG |
K622R |
NP_001252507.1 |
NM_001265579.1 |
4805 |
Missense Mutation |
AAG,AGG |
K608R |
NP_001252508.1 |
NM_001265580.1 |
4805 |
Missense Mutation |
AAG,AGG |
K608R |
NP_001252509.1 |
NM_014883.3 |
4805 |
Missense Mutation |
AAG,AGG |
K962R |
NP_055698.2 |
XM_005262681.2 |
4805 |
Missense Mutation |
AAG,AGG |
K948R |
XP_005262738.1 |
XM_005262682.2 |
4805 |
Missense Mutation |
AAG,AGG |
K942R |
XP_005262739.1 |
XM_005262683.2 |
4805 |
Missense Mutation |
AAG,AGG |
K934R |
XP_005262740.1 |
XM_005262684.3 |
4805 |
Missense Mutation |
AAG,AGG |
K753R |
XP_005262741.1 |
XM_006714057.3 |
4805 |
Missense Mutation |
AAG,AGG |
K773R |
XP_006714120.1 |
XM_011531516.1 |
4805 |
Missense Mutation |
AAG,AGG |
K962R |
XP_011529818.1 |
XM_011531517.1 |
4805 |
Missense Mutation |
AAG,AGG |
K934R |
XP_011529819.1 |
XM_011531518.1 |
4805 |
Missense Mutation |
AAG,AGG |
K776R |
XP_011529820.1 |
XM_011531519.2 |
4805 |
Missense Mutation |
AAG,AGG |
K776R |
XP_011529821.1 |
XM_017007624.1 |
4805 |
Missense Mutation |
AAG,AGG |
K920R |
XP_016863113.1 |
XM_017007625.1 |
4805 |
Missense Mutation |
AAG,AGG |
K907R |
XP_016863114.1 |
XM_017007626.1 |
4805 |
Missense Mutation |
AAG,AGG |
K785R |
XP_016863115.1 |
XM_017007627.1 |
4805 |
Missense Mutation |
AAG,AGG |
K753R |
XP_016863116.1 |
XM_017007628.1 |
4805 |
Missense Mutation |
AAG,AGG |
K711R |
XP_016863117.1 |
XM_017007629.1 |
4805 |
Missense Mutation |
AAG,AGG |
K690R |
XP_016863118.1 |
XM_017007630.1 |
4805 |
Missense Mutation |
AAG,AGG |
K690R |
XP_016863119.1 |
XM_017007631.1 |
4805 |
Missense Mutation |
AAG,AGG |
K676R |
XP_016863120.1 |
XM_017007632.1 |
4805 |
Missense Mutation |
AAG,AGG |
K676R |
XP_016863121.1 |
XM_017007633.1 |
4805 |
Missense Mutation |
AAG,AGG |
K641R |
XP_016863122.1 |
XM_017007634.1 |
4805 |
Missense Mutation |
AAG,AGG |
K594R |
XP_016863123.1 |
- Gene
- FAM13A-AS1
- Gene Name
- FAM13A antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details