Product Details

SNP ID
rs202196466
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:185145802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAACGTGCACATTTTTGTGAGCTG[C/G]ATGATTGCCCAGAGTGTGACGGCAG
Phenotype
MIM: 103220
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC25A4 PubMed Links

Gene Details

Gene
SLC25A4
Gene Name
solute carrier family 25 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001151.3 752 Missense Mutation TGC,TGG C214W NP_001142.2

View Full Product Details