Product Details
- SNP ID
-
rs202233404
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:185399743 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGACAGAAACGGAGTCTCGGAAGTG[C/T]AGAAAATACCAGTGGGAAAAGGAAG
- Phenotype
-
MIM: 611482
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ANKRD37
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4317244] are located under a probe and SNP(s) [rs4631093] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ANKRD37
- Gene Name
- ankyrin repeat domain 37
- Gene
- C4orf47
- Gene Name
- chromosome 4 open reading frame 47
There are no transcripts associated with this gene.
- Gene
- LRP2BP
- Gene Name
- LRP2 binding protein
There are no transcripts associated with this gene.
- Gene
- UFSP2
- Gene Name
- UFM1 specific peptidase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018359.3 |
2176 |
UTR 3 |
|
|
NP_060829.2 |
View Full Product Details