Product Details
- SNP ID
-
rs202133904
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:10973625 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGGACCTGGTCCTCGAAGAAGGAC[C/T]CATCGTAATTTCTTGTGGAATTCTG
- Phenotype
-
MIM: 604275
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CTNND2
PubMed Links
Gene Details
- Gene
- CTNND2
- Gene Name
- catenin delta 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001288715.1 |
3486 |
Missense Mutation |
GAG,GGG |
E1078G |
NP_001275644.1 |
NM_001288716.1 |
3486 |
Missense Mutation |
GAG,GGG |
E832G |
NP_001275645.1 |
NM_001288717.1 |
3486 |
Missense Mutation |
GAG,GGG |
E736G |
NP_001275646.1 |
NM_001332.3 |
3486 |
Missense Mutation |
GAG,GGG |
E1169G |
NP_001323.1 |
XM_005248251.3 |
3486 |
Missense Mutation |
GAG,GGG |
E1194G |
XP_005248308.1 |
XM_005248252.2 |
3486 |
Missense Mutation |
GAG,GGG |
E1180G |
XP_005248309.1 |
XM_005248253.1 |
3486 |
Missense Mutation |
GAG,GGG |
E1103G |
XP_005248310.1 |
XM_011513967.2 |
3486 |
Missense Mutation |
GAG,GGG |
E1103G |
XP_011512269.1 |
XM_017009072.1 |
3486 |
Missense Mutation |
GAG,GGG |
E948G |
XP_016864561.1 |
XM_017009073.1 |
3486 |
Missense Mutation |
GAG,GGG |
E934G |
XP_016864562.1 |
XM_017009074.1 |
3486 |
Missense Mutation |
GAG,GGG |
E923G |
XP_016864563.1 |
XM_017009075.1 |
3486 |
Missense Mutation |
GAG,GGG |
E857G |
XP_016864564.1 |
XM_017009076.1 |
3486 |
Missense Mutation |
GAG,GGG |
E857G |
XP_016864565.1 |
View Full Product Details