Product Details

SNP ID
rs201641885
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:134661343 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCATAGTGGTCCTGCTCCCCGA[A/G]TGCCATTACCTGCTTCTCAGAACCC
Phenotype
MIM: 607183
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEC24A PubMed Links

Gene Details

Gene
SEC24A
Gene Name
SEC24 homolog A, COPII coat complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252231.1 259 Missense Mutation ATG,GTG M108V NP_001239160.1
NM_021982.2 259 Missense Mutation ATG,GTG M108V NP_068817.1
XM_006714523.2 259 Missense Mutation ATG,GTG M108V XP_006714586.1
XM_017008961.1 259 Missense Mutation ATG,GTG M82V XP_016864450.1
XM_017008962.1 259 Intron XP_016864451.1
XM_017008963.1 259 Missense Mutation ATG,GTG M108V XP_016864452.1

View Full Product Details