Product Details

SNP ID
rs199734683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:151505738 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCATACCCACCCCCTTGTAGCCCC[C/T]GTCTGCCAGGCAGGGCCCTCCCCCT
Phenotype
MIM: 604269 MIM: 606561
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAT2 PubMed Links

Gene Details

Gene
FAT2
Gene Name
FAT atypical cadherin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001447.2 13231 Missense Mutation AGG,GGG R4293G NP_001438.1
XM_006714761.3 13231 Missense Mutation AGG,GGG R4293G XP_006714824.1
XM_011537600.2 13231 Missense Mutation AGG,GGG R4293G XP_011535902.1
XM_011537603.2 13231 Missense Mutation AGG,GGG R4293G XP_011535905.1
XM_017009224.1 13231 Missense Mutation AGG,GGG R4293G XP_016864713.1
XM_017009225.1 13231 Missense Mutation AGG,GGG R4293G XP_016864714.1
XM_017009226.1 13231 Intron XP_016864715.1
XM_017009227.1 13231 Intron XP_016864716.1
Gene
SLC36A1
Gene Name
solute carrier family 36 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308150.1 13231 Intron NP_001295079.1
NM_001308151.1 13231 Intron NP_001295080.1
NM_078483.3 13231 Intron NP_510968.2
XM_005268386.1 13231 Intron XP_005268443.1
XM_006714759.3 13231 Intron XP_006714822.1
XM_011537580.2 13231 Intron XP_011535882.1
XM_011537581.1 13231 Intron XP_011535883.1
XM_011537583.2 13231 Intron XP_011535885.1
XM_011537584.2 13231 Intron XP_011535886.1
XM_011537585.1 13231 Intron XP_011535887.1
XM_011537586.2 13231 Intron XP_011535888.1
XM_011537587.2 13231 Intron XP_011535889.1
XM_011537588.2 13231 Intron XP_011535890.1
XM_011537589.2 13231 Intron XP_011535891.1
XM_011537590.1 13231 Intron XP_011535892.1
XM_011537591.1 13231 Intron XP_011535893.1
XM_011537592.2 13231 Intron XP_011535894.1
XM_011537593.2 13231 Intron XP_011535895.1
XM_011537594.1 13231 Intron XP_011535896.1
XM_011537595.2 13231 Intron XP_011535897.2
XM_011537596.2 13231 Intron XP_011535898.2
XM_017009216.1 13231 Intron XP_016864705.1
XM_017009217.1 13231 Intron XP_016864706.1
XM_017009218.1 13231 Intron XP_016864707.1
XM_017009219.1 13231 Intron XP_016864708.1
XM_017009220.1 13231 Intron XP_016864709.1

View Full Product Details