Product Details

SNP ID
rs199818174
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:152392337 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGTCAGCTCCACAAAGTGGCATT[C/G]TGTCAGGAAGATGTTCCGCTGGGCA
Phenotype
MIM: 605108
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NMUR2 PubMed Links

Gene Details

Gene
NMUR2
Gene Name
neuromedin U receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020167.4 1268 Missense Mutation CAA,GAA Q368E NP_064552.3
XM_011537670.2 1268 Intron XP_011535972.1

View Full Product Details