Product Details

SNP ID
rs199749596
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:81304867 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGAGGGAACAAGCCAAAACCTAC[C/G]AAAGCGTAAACAACTTGAAGCCAAT
Phenotype
MIM: 614315
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ACOT12 PubMed Links

Gene Details

Gene
ACOT12
Gene Name
acyl-CoA thioesterase 12
There are no transcripts associated with this gene.

Gene
CKMT2-AS1
Gene Name
CKMT2 antisense RNA 1
There are no transcripts associated with this gene.

Gene
ZCCHC9
Gene Name
zinc finger CCHC-type containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001131035.1 600 Missense Mutation CCA,CGA P37R NP_001124507.1
NM_001131036.1 600 Missense Mutation CCA,CGA P37R NP_001124508.1
NM_032280.2 600 Missense Mutation CCA,CGA P37R NP_115656.1
XM_005248616.3 600 Intron XP_005248673.1

View Full Product Details