Product Details

SNP ID
rs201600677
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:171311582 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGATAGTTCCAAGGTTCCCGCT[C/G]TCACCTCCCTGGTGTGAGCCCACCA
Phenotype
MIM: 604640
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC728145 PubMed Links

Gene Details

Gene
LOC728145
Gene Name
collagen alpha-1(I) chain
There are no transcripts associated with this gene.

Gene
TLX3
Gene Name
T-cell leukemia homeobox 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021025.3 941 Missense Mutation CTC,GTC L287V NP_066305.2

View Full Product Details