Product Details

SNP ID
rs200417568
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:177403357 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGCTGAACGTAAGGCGACAGGAG[C/T]GCGCAGCTGCCGTCCGCATCCTCCT
Phenotype
MIM: 610619 MIM: 612812 MIM: 182309
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
F12 PubMed Links

Gene Details

Gene
F12
Gene Name
coagulation factor XII
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000505.3 1477 Silent Mutation GCA,GCG A476A NP_000496.2
XM_011534462.2 1477 Silent Mutation GCA,GCG A364A XP_011532764.1
Gene
PFN3
Gene Name
profilin 3
There are no transcripts associated with this gene.

Gene
SLC34A1
Gene Name
solute carrier family 34 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167579.1 1477 Intron NP_001161051.1
NM_003052.4 1477 Intron NP_003043.3
XM_005265975.1 1477 Intron XP_005266032.1
XM_017009773.1 1477 Intron XP_016865262.1
XM_017009774.1 1477 Intron XP_016865263.1
XM_017009775.1 1477 Intron XP_016865264.1

View Full Product Details