Product Details
- SNP ID
-
rs201229460
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:38942332 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGATTCAGCAGATGTATCAACTATA[A/G]GTTGCTTTGGTGGTGTTGCCAACAC
- Phenotype
-
MIM: 601743
MIM: 609022
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OSMR
PubMed Links
Gene Details
- Gene
- OSMR
- Gene Name
- oncostatin M receptor
- Gene
- RICTOR
- Gene Name
- RPTOR independent companion of MTOR complex 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001285439.1 |
5014 |
Missense Mutation |
CCT,CTT |
P1724L |
NP_001272368.1 |
NM_001285440.1 |
5014 |
Missense Mutation |
CCT,CTT |
P1377L |
NP_001272369.1 |
NM_152756.4 |
5014 |
Missense Mutation |
CCT,CTT |
P1700L |
NP_689969.2 |
XM_006714463.3 |
5014 |
Missense Mutation |
CCT,CTT |
P1662L |
XP_006714526.1 |
XM_011514005.2 |
5014 |
Missense Mutation |
CCT,CTT |
P1686L |
XP_011512307.1 |
XM_011514006.2 |
5014 |
Missense Mutation |
CCT,CTT |
P1661L |
XP_011512308.1 |
XM_017009311.1 |
5014 |
Missense Mutation |
CCT,CTT |
P1708L |
XP_016864800.1 |
XM_017009312.1 |
5014 |
Missense Mutation |
CCT,CTT |
P1684L |
XP_016864801.1 |
XM_017009313.1 |
5014 |
Missense Mutation |
CCT,CTT |
P1671L |
XP_016864802.1 |
XM_017009314.1 |
5014 |
Missense Mutation |
CCT,CTT |
P1439L |
XP_016864803.1 |
XM_017009315.1 |
5014 |
Missense Mutation |
CCT,CTT |
P1415L |
XP_016864804.1 |
View Full Product Details