Product Details

SNP ID
rs201231990
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:75029220 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCTTCACATATTCATAAGGCACC[C/T]GTCTAAGTTCATGATGGTAAGTGAA
Phenotype
MIM: 616782
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GCNT4 PubMed Links

Gene Details

Gene
GCNT4
Gene Name
glucosaminyl (N-acetyl) transferase 4, core 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016591.2 1680 Missense Mutation CAG,CGG Q273R NP_057675.1

View Full Product Details