Product Details

SNP ID
rs200760299
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27451432 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGTGCTGAATGAGATATGTGCT[C/G]TGGCTAAAAGTCTTTCTGCATTCAT
Phenotype
MIM: 602277
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
ZNF184 PubMed Links

Gene Details

Gene
ZNF184
Gene Name
zinc finger protein 184
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318891.1 2412 Missense Mutation CAC,CAG H709Q NP_001305820.1
NM_001318892.1 2412 Missense Mutation CAC,CAG H709Q NP_001305821.1
NM_001318893.1 2412 Missense Mutation CAC,CAG H633Q NP_001305822.1
NM_007149.2 2412 Missense Mutation CAC,CAG H709Q NP_009080.2
XM_011514861.2 2412 Missense Mutation CAC,CAG H633Q XP_011513163.1

View Full Product Details