Product Details

SNP ID
rs201573534
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:110392988 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCCTCAAGCCCACCTTCTCCCT[G/T]ATGTTGCAGGCTCCGTGGAGGGAGG
Phenotype
MIM: 124450
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DDO PubMed Links

Gene Details

Gene
DDO
Gene Name
D-aspartate oxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003649.2 724 Silent Mutation ATA,ATC I299I NP_003640.2
NM_004032.2 724 Silent Mutation ATA,ATC I240I NP_004023.2

View Full Product Details