Product Details

SNP ID
rs200863978
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29486787 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATTATGTTTCCACATCGACCCT[A/G]TGCTCCCTGGGAAGAAGGTTCTCCA
Phenotype
MIM: 607235
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAS1L PubMed Links

Gene Details

Gene
MAS1L
Gene Name
MAS1 proto-oncogene like, G protein-coupled receptor
There are no transcripts associated with this gene.

View Full Product Details