Product Details

SNP ID
rs200883410
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:82365409 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCAGCCTGCGCCAGCTCGACCTC[A/T]GCCACAACCCACTGGCCGACCTCAG
Phenotype
MIM: 190920
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TPBG PubMed Links

Gene Details

Gene
TPBG
Gene Name
trophoblast glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166392.1 873 Missense Mutation AGC,TGC S150C NP_001159864.1
NM_006670.4 873 Missense Mutation AGC,TGC S150C NP_006661.1
XM_011536097.2 873 Missense Mutation AGC,TGC S150C XP_011534399.1

View Full Product Details