Product Details
- SNP ID
-
rs201535868
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:49786169 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAGGCATCCCATTCAAATACTCCT[A/G]ACGGCCCATTCCAAACAATTAGCCT
- Phenotype
-
MIM: 172270
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PGK2
PubMed Links
Gene Details
- Gene
- PGK2
- Gene Name
- phosphoglycerate kinase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_138733.4 |
1172 |
Missense Mutation |
TCA,TTA |
S340L |
NP_620061.2 |
View Full Product Details