Product Details

SNP ID
rs200553902
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:55174701 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCCACCGACTATGACGACGAGGA[A/G]TTCCTGCGGTACCTGTGGAGGGAAT
Phenotype
MIM: 602393
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HCRTR2 PubMed Links

Gene Details

Gene
HCRTR2
Gene Name
hypocretin receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001526.3 429 Silent Mutation GAA,GAG E38E NP_001517.2
XM_017010798.1 429 Silent Mutation GAA,GAG E38E XP_016866287.1

View Full Product Details