Product Details

SNP ID
rs200166268
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27807640 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTCTGGATCTCCCTGGAGGTGAT[C/G]GTCGAGCGCTTGTTGTAGTGCGCCA
Phenotype
MIM: 602787 MIM: 602791 MIM: 602800 MIM: 602802 MIM: 602818
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HIST1H2AI PubMed Links

Gene Details

Gene
HIST1H2AI
Gene Name
histone cluster 1, H2ai
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003509.2 Intron NP_003500.1
Gene
HIST1H2AJ
Gene Name
histone cluster 1, H2aj
There are no transcripts associated with this gene.

Gene
HIST1H2BL
Gene Name
histone cluster 1, H2bl
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003519.3 Intron NP_003510.1
Gene
HIST1H2BM
Gene Name
histone cluster 1, H2bm
There are no transcripts associated with this gene.

Gene
HIST1H3H
Gene Name
histone cluster 1, H3h
There are no transcripts associated with this gene.

View Full Product Details