Product Details

SNP ID
rs199643288
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27451811 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACTCATTACACTTGTAAGGTCGT[C/T]CTCCAGTATGGATCTTCCTATGCTG
Phenotype
MIM: 602277
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF184 PubMed Links

Gene Details

Gene
ZNF184
Gene Name
zinc finger protein 184
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318891.1 2033 Missense Mutation GAA,GGA E583G NP_001305820.1
NM_001318892.1 2033 Missense Mutation GAA,GGA E583G NP_001305821.1
NM_001318893.1 2033 Missense Mutation GAA,GGA E507G NP_001305822.1
NM_007149.2 2033 Missense Mutation GAA,GGA E583G NP_009080.2
XM_011514861.2 2033 Missense Mutation GAA,GGA E507G XP_011513163.1

View Full Product Details