Product Details
- SNP ID
-
rs199723665
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:3154847 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCTCTCTGACTCCTTCCTCACCAC[A/G]TCCAGGACCGAGTCGACCAGCTCGG
- Phenotype
-
MIM: 603156
MIM: 615101
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BPHL
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs143644817] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BPHL
- Gene Name
- biphenyl hydrolase-like (serine hydrolase)
There are no transcripts associated with this gene.
- Gene
- TUBB2A
- Gene Name
- tubulin beta 2A class IIa
View Full Product Details