Product Details

SNP ID
rs200314349
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29086614 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGCCATAGGAGATGAGGATCAAT[C/G]TCACTGGAATTAGAAGAATTAGTAC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC100129636 PubMed Links

Gene Details

Gene
LOC100129636
Gene Name
uncharacterized LOC100129636
There are no transcripts associated with this gene.

Gene
OR2B3
Gene Name
olfactory receptor family 2 subfamily B member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005226.2 700 Missense Mutation ACA,AGA T212R NP_001005226.1

View Full Product Details