Product Details

SNP ID
rs200726414
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:156950266 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTGCCCGGAAACGGGCGCTTTTA[A/G]CGGCGAACGAGGAGGAGGACCGAGA
Phenotype
MIM: 611269
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NOM1 PubMed Links

Gene Details

Gene
NOM1
Gene Name
nucleolar protein with MIF4G domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138400.1 544 Missense Mutation ACG,GCG T177A NP_612409.1

View Full Product Details