Product Details

SNP ID
rs200748640
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:27143329 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCGGATCGGGCTGAGGAGAGTG[C/G]GTGGACGTGGCCGGCTGGCTGTACC
Phenotype
MIM: 142954 MIM: 142952 MIM: 142951
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HOXA-AS3 PubMed Links

Gene Details

Gene
HOXA-AS3
Gene Name
HOXA cluster antisense RNA 3
There are no transcripts associated with this gene.

Gene
HOXA3
Gene Name
homeobox A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030661.4 340 Intron NP_109377.1
NM_153631.2 340 Intron NP_705895.1
XM_005249730.2 340 Intron XP_005249787.1
XM_005249731.2 340 Intron XP_005249788.1
XM_005249732.3 340 Intron XP_005249789.1
XM_006715715.2 340 Intron XP_006715778.1
XM_011515343.2 340 Intron XP_011513645.1
Gene
HOXA5
Gene Name
homeobox A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019102.3 340 Silent Mutation ACC,ACG T93T NP_061975.2
Gene
HOXA6
Gene Name
homeobox A6
There are no transcripts associated with this gene.

View Full Product Details