Product Details

SNP ID
rs199922834
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:96694799 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAAAATCTGGCTTAAAACTTACC[A/G]TAACTGATTAGAGAAGTCATCCTCT
Phenotype
MIM: 601285
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SHFM1 PubMed Links

Gene Details

Gene
SHFM1
Gene Name
split hand/foot malformation (ectrodactyly) type 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006304.1 Intron NP_006295.1

View Full Product Details