Product Details

SNP ID
rs199923658
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:128715578 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTCTTGCCTGTGGATGGAGAGC[C/T]GAACCCTGGAGCGGGCCTAGGTGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM71F1 PubMed Links

Gene Details

Gene
FAM71F1
Gene Name
family with sequence similarity 71 member F1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282788.1 280 Missense Mutation CCG,CTG P46L NP_001269717.1
NM_001282789.1 280 Intron NP_001269718.1
NM_032599.3 280 Missense Mutation CCG,CTG P46L NP_115988.1
XM_017012743.1 280 Missense Mutation CCG,CTG P46L XP_016868232.1

View Full Product Details