Product Details
- SNP ID
-
rs200091728
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:77312128 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTTCTTATAGTTCTGAAGCAGTCG[C/T]GTCACGTGGTTCCGCGAAATGATGT
- Phenotype
-
MIM: 613552
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GSAP
PubMed Links
Gene Details
- Gene
- GSAP
- Gene Name
- gamma-secretase activating protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017439.3 |
2438 |
Silent Mutation |
ACA,ACG |
T782T |
NP_059135.2 |
XM_011516325.2 |
2438 |
Silent Mutation |
ACA,ACG |
T800T |
XP_011514627.1 |
XM_011516326.2 |
2438 |
Silent Mutation |
ACA,ACG |
T782T |
XP_011514628.1 |
XM_011516327.2 |
2438 |
Silent Mutation |
ACA,ACG |
T800T |
XP_011514629.1 |
XM_011516330.2 |
2438 |
Silent Mutation |
ACA,ACG |
T585T |
XP_011514632.1 |
XM_011516331.2 |
2438 |
Intron |
|
|
XP_011514633.1 |
XM_011516332.2 |
2438 |
Intron |
|
|
XP_011514634.1 |
XM_017012343.1 |
2438 |
Silent Mutation |
ACA,ACG |
T768T |
XP_016867832.1 |
XM_017012344.1 |
2438 |
Silent Mutation |
ACA,ACG |
T764T |
XP_016867833.1 |
XM_017012345.1 |
2438 |
Silent Mutation |
ACA,ACG |
T782T |
XP_016867834.1 |
XM_017012346.1 |
2438 |
Silent Mutation |
ACA,ACG |
T585T |
XP_016867835.1 |
XM_017012347.1 |
2438 |
Silent Mutation |
ACA,ACG |
T535T |
XP_016867836.1 |
XM_017012348.1 |
2438 |
Silent Mutation |
ACA,ACG |
T585T |
XP_016867837.1 |
XM_017012349.1 |
2438 |
Silent Mutation |
ACA,ACG |
T585T |
XP_016867838.1 |
XM_017012350.1 |
2438 |
Intron |
|
|
XP_016867839.1 |
XM_017012351.1 |
2438 |
Intron |
|
|
XP_016867840.1 |
View Full Product Details