Product Details

SNP ID
rs202145310
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:75811886 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGGGAAGCCTTCTTCTGCTTGGC[A/G]TCCAGGTTCTTCATGTACCTCTGGA
Phenotype
MIM: 602495
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCL24 PubMed Links

Gene Details

Gene
CCL24
Gene Name
C-C motif chemokine ligand 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002991.2 537 Silent Mutation GAC,GAT D90D NP_002982.2
XM_011516459.2 537 Silent Mutation GAC,GAT D90D XP_011514761.1
XM_011516460.2 537 Silent Mutation GAC,GAT D90D XP_011514762.1

View Full Product Details