Product Details

SNP ID
rs202182643
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:30019116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAATATCAGTGTACTTTCTGGGGG[A/C]ATTTTACCTGACGTGAGGAAAGAAG
Phenotype
MIM: 614505 MIM: 608639
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FKBP14 PubMed Links

Gene Details

Gene
FKBP14
Gene Name
FK506 binding protein 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017946.3 650 Missense Mutation ATG,ATT M119I NP_060416.1
Gene
PLEKHA8
Gene Name
pleckstrin homology domain containing A8
There are no transcripts associated with this gene.

View Full Product Details