Product Details

SNP ID
rs201100435
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99400377 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTTTGGTTGTCTGTGCCACCCG[A/G]TTGGGGTTCTCGATGTCGATGAGCC
Phenotype
MIM: 604223 MIM: 603477 MIM: 607075
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARPC1B PubMed Links

Gene Details

Gene
ARPC1B
Gene Name
actin related protein 2/3 complex subunit 1B
There are no transcripts associated with this gene.

Gene
BUD31
Gene Name
BUD31 homolog
There are no transcripts associated with this gene.

Gene
PDAP1
Gene Name
PDGFA associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014891.6 395 Silent Mutation AAC,AAT N87N NP_055706.1

View Full Product Details