Product Details

SNP ID
rs201327269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:149003992 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTTGGCAGTGGCGTCCATCTTGGC[A/G]ATGACCAGGCCCTTTTGGCCCTTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDIA4 PubMed Links

Gene Details

Gene
PDIA4
Gene Name
protein disulfide isomerase family A member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004911.4 2022 Silent Mutation ATC,ATT I580I NP_004902.1
XM_006716185.2 2022 Silent Mutation ATC,ATT I581I XP_006716248.1

View Full Product Details