Product Details

SNP ID
rs201871107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:75422005 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCAGGCACGATCTTGATCGTGGA[C/T]GCAGGTGCAGGTGTGGGTGCAGTAG
Phenotype
MIM: 615754
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NSUN5P1 PubMed Links

Gene Details

Gene
NSUN5P1
Gene Name
NOP2/Sun RNA methyltransferase family member 5 pseudogene 1
There are no transcripts associated with this gene.

Gene
POM121C
Gene Name
POM121 transmembrane nucleoporin C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099415.2 Intron NP_001092885.2

View Full Product Details