Product Details

SNP ID
rs199541377
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:103103532 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATCTCTCTAGAGCTTCATTCAGC[C/T]TCACTGGAGGCTCTAAGTAATGCTG
Phenotype
MIM: 611864 MIM: 612334
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARMC10 PubMed Links

Gene Details

Gene
ARMC10
Gene Name
armadillo repeat containing 10
There are no transcripts associated with this gene.

Gene
NAPEPLD
Gene Name
N-acyl phosphatidylethanolamine phospholipase D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122838.1 1407 Missense Mutation AAG,AGG K360R NP_001116310.1
NM_198990.4 1407 Missense Mutation AAG,AGG K360R NP_945341.3
XM_005250214.4 1407 Missense Mutation AAG,AGG K360R XP_005250271.2
XM_005250215.2 1407 Missense Mutation AAG,AGG K360R XP_005250272.1
XM_005250218.2 1407 Missense Mutation AAG,AGG K360R XP_005250275.1
XM_011515933.2 1407 Missense Mutation AAG,AGG K360R XP_011514235.1
XM_011515934.2 1407 Missense Mutation AAG,AGG K225R XP_011514236.1
XM_017011854.1 1407 Missense Mutation AAG,AGG K360R XP_016867343.1
XM_017011855.1 1407 Missense Mutation AAG,AGG K360R XP_016867344.1
XM_017011856.1 1407 Intron XP_016867345.1
XM_017011857.1 1407 Intron XP_016867346.1

View Full Product Details