Product Details

SNP ID
rs201510307
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144881750 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCAGGCTGAAGGCTTTGCCACAT[G/T]CTGTGCACTGGAAGGGCTTTTCACC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF250 PubMed Links

Gene Details

Gene
ZNF250
Gene Name
zinc finger protein 250
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109689.3 1565 Missense Mutation GAA,GCA E478A NP_001103159.1
NM_021061.4 1565 Missense Mutation GAA,GCA E483A NP_066405.1
XM_005272327.1 1565 Missense Mutation GAA,GCA E478A XP_005272384.1
XM_005272328.3 1565 Missense Mutation GAA,GCA E478A XP_005272385.1
XM_006716612.3 1565 Missense Mutation GAA,GCA E483A XP_006716675.1
XM_006716613.3 1565 Missense Mutation GAA,GCA E483A XP_006716676.1
XM_011517209.1 1565 Missense Mutation GAA,GCA E478A XP_011515511.1

View Full Product Details