Product Details

SNP ID
rs199617556
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:86048652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCTTCATGGAAGACCCTGCAGCC[A/G]TGGTGATCACCCAGGGATGGTCCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PSKH2 PubMed Links

Gene Details

Gene
PSKH2
Gene Name
protein serine kinase H2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033126.1 968 Missense Mutation ACG,ATG T323M NP_149117.1
XM_017013929.1 968 Missense Mutation ACG,ATG T442M XP_016869418.1

View Full Product Details