Product Details

SNP ID
rs200470486
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144317443 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGAGCCAGATGAGGTGATTGGG[A/G]ACCTGGCAGGGAGGTGGGGGTGGGC
Phenotype
MIM: 604900 MIM: 140580
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DGAT1 PubMed Links

Gene Details

Gene
DGAT1
Gene Name
diacylglycerol O-acyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012079.5 988 Silent Mutation GTC,GTT V328V NP_036211.2
XM_011517356.2 988 Silent Mutation GTC,GTT V271V XP_011515658.1
Gene
HSF1
Gene Name
heat shock transcription factor 1
There are no transcripts associated with this gene.

Gene
MIR6848
Gene Name
microRNA 6848
There are no transcripts associated with this gene.

View Full Product Details