Product Details

SNP ID
rs200110154
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:38267227 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTTCTAGGTTAAATTCAATGATG[C/T]GAAAACAAGCATAGGGTAGAGTCCA
Phenotype
MIM: 615003 MIM: 610626 MIM: 607083
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDHD2 PubMed Links

Gene Details

Gene
DDHD2
Gene Name
DDHD domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164232.1 1190 Intron NP_001157704.1
NM_001164234.1 1190 Intron NP_001157706.1
NM_015214.2 1190 Intron NP_056029.2
XM_005273454.2 1190 Intron XP_005273511.1
XM_005273455.3 1190 Intron XP_005273512.1
XM_005273456.3 1190 Intron XP_005273513.1
XM_011544456.1 1190 Intron XP_011542758.1
XM_017013252.1 1190 Intron XP_016868741.1
XM_017013253.1 1190 Intron XP_016868742.1
XM_017013254.1 1190 Intron XP_016868743.1
XM_017013255.1 1190 Intron XP_016868744.1
XM_017013256.1 1190 Intron XP_016868745.1
Gene
PLPP5
Gene Name
phospholipid phosphatase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102559.1 1190 Intron NP_001096029.1
NM_001102560.1 1190 Missense Mutation CAC,CGC H168R NP_001096030.1
NM_032483.3 1190 Intron NP_115872.2
XM_005273661.4 1190 Intron XP_005273718.2
XM_005273663.3 1190 UTR 3 XP_005273720.2
XM_011544670.2 1190 UTR 3 XP_011542972.1
XM_011544671.1 1190 Intron XP_011542973.1
XM_011544672.1 1190 Intron XP_011542974.1
XM_011544674.2 1190 UTR 3 XP_011542976.1
XM_011544675.2 1190 UTR 3 XP_011542977.1
XM_017013905.1 1190 UTR 3 XP_016869394.1
XM_017013906.1 1190 UTR 3 XP_016869395.1
XM_017013907.1 1190 Intron XP_016869396.1
Gene
WHSC1L1
Gene Name
Wolf-Hirschhorn syndrome candidate 1-like 1
There are no transcripts associated with this gene.

View Full Product Details