Product Details

SNP ID
rs200563412
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:133038691 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAAGGCCATAGCTGAAAAGGGACT[C/T]GTCTACCCCAAGCGGGTTCTCTGTT
Phenotype
MIM: 601099 MIM: 188450
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR7848 PubMed Links

Gene Details

Gene
MIR7848
Gene Name
microRNA 7848
There are no transcripts associated with this gene.

Gene
SLA
Gene Name
Src-like-adaptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001045556.2 1086 Missense Mutation AAG,GAG K222E NP_001039021.1
NM_001045557.2 1086 Missense Mutation AAG,GAG K239E NP_001039022.2
NM_001282964.1 1086 Missense Mutation AAG,GAG K195E NP_001269893.1
NM_001282965.1 1086 Missense Mutation AAG,GAG K114E NP_001269894.1
NM_006748.3 1086 Missense Mutation AAG,GAG K262E NP_006739.2
XM_017013739.1 1086 Missense Mutation AAG,GAG K114E XP_016869228.1
Gene
TG
Gene Name
thyroglobulin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003235.4 1086 Intron NP_003226.4
XM_005251038.4 1086 Intron XP_005251095.1
XM_005251040.4 1086 Intron XP_005251097.1
XM_005251042.4 1086 Intron XP_005251099.1
XM_006716622.3 1086 Intron XP_006716685.1
XM_017013793.1 1086 Intron XP_016869282.1
XM_017013794.1 1086 Intron XP_016869283.1
XM_017013795.1 1086 Intron XP_016869284.1
XM_017013796.1 1086 Intron XP_016869285.1
XM_017013797.1 1086 Intron XP_016869286.1
XM_017013798.1 1086 Intron XP_016869287.1
XM_017013799.1 1086 Intron XP_016869288.1
XM_017013800.1 1086 Intron XP_016869289.1

View Full Product Details