Product Details

SNP ID
rs201453879
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127868063 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGAGAAGACACTGTCCACGGAG[C/T]CCTCAGCGTTGACCTGTGGGGAGAT
Phenotype
MIM: 103000
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AK1 PubMed Links

Gene Details

Gene
AK1
Gene Name
adenylate kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000476.2 1030 Missense Mutation NP_000467.1
NM_001318121.1 1030 Missense Mutation NP_001305050.1
NM_001318122.1 1030 Missense Mutation NP_001305051.1
XM_017014427.1 1030 Missense Mutation XP_016869916.1
XM_017014428.1 1030 Missense Mutation XP_016869917.1
Gene
MIR4672
Gene Name
microRNA 4672
There are no transcripts associated with this gene.

View Full Product Details