Product Details

SNP ID
rs200259060
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136362175 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTCTGGCCGTCAGGATCTCTTCG[A/G]TATTTCTGGGGGGCAGGGAGGCAAC
Phenotype
MIM: 607212 MIM: 609491
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CARD9 PubMed Links

Gene Details

Gene
CARD9
Gene Name
caspase recruitment domain family member 9
There are no transcripts associated with this gene.

Gene
DNLZ
Gene Name
DNL-type zinc finger
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080849.2 471 Missense Mutation ACC,ATC T125I NP_001074318.1
Gene
GPSM1
Gene Name
G-protein signaling modulator 1
There are no transcripts associated with this gene.

View Full Product Details