Product Details

SNP ID
rs200016170
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137009802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGGCTGGGTACCCAATGAGGGC[A/G]ATGGCCGTGGAGAGCTTCCGCTTGT
Phenotype
MIM: 600047
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCA2 PubMed Links

Gene Details

Gene
ABCA2
Gene Name
ATP binding cassette subfamily A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001606.4 6679 Silent Mutation ATC,ATT I2199I NP_001597.2
NM_212533.2 6679 Silent Mutation ATC,ATT I2229I NP_997698.1
XM_006716996.3 6679 Silent Mutation ATC,ATT I2198I XP_006717059.1

View Full Product Details