Product Details

SNP ID
rs199959705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128792574 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTGCAGATCGACAAAGATGTCC[A/G]GTAGGCAGGGTGCCTGGGGCCGGGA
Phenotype
MIM: 616218
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TBC1D13 PubMed Links

Gene Details

Gene
TBC1D13
Gene Name
TBC1 domain family member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286772.1 433 Missense Mutation CAG,CGG Q128R NP_001273701.1
NM_018201.4 433 Missense Mutation CAG,CGG Q128R NP_060671.3
XM_005252060.2 433 Missense Mutation CAG,CGG Q52R XP_005252117.1

View Full Product Details