Product Details

SNP ID
rs199614285
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:108934516 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACCGAGCCCGAAGCCGCGGAGCC[C/G]GTGGTGCCTTCGCTCGTGGATCGAT
Phenotype
MIM: 604785 MIM: 603722
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
CTNNAL1 PubMed Links
Additional Information
For this assay, SNP(s) [rs76817627] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CTNNAL1
Gene Name
catenin alpha like 1
There are no transcripts associated with this gene.

Gene
FAM206A
Gene Name
family with sequence similarity 206 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017832.3 120 Silent Mutation CCC,CCG P10P NP_060302.1
XM_011518816.2 120 Silent Mutation CCC,CCG P10P XP_011517118.1
Gene
IKBKAP
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318360.1 120 Intron NP_001305289.1
NM_003640.4 120 Intron NP_003631.2
XM_011519136.1 120 Intron XP_011517438.1
XM_011519137.1 120 Intron XP_011517439.1
XM_017015238.1 120 Intron XP_016870727.1

View Full Product Details