Product Details

SNP ID
rs200049812
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128706778 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGCCAGCTGAAGGTGGCCCTGCT[G/T]CGGATGAAGATCAGCAGCCTGGAGG
Phenotype
MIM: 610714
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PKN3 PubMed Links

Gene Details

Gene
PKN3
Gene Name
protein kinase N3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317926.1 960 Silent Mutation CTG,CTT L159L NP_001304855.1
NM_013355.4 960 Silent Mutation CTG,CTT L159L NP_037487.2
XM_005251946.3 960 Silent Mutation CTG,CTT L156L XP_005252003.1
XM_006717080.2 960 Silent Mutation CTG,CTT L166L XP_006717143.1
XM_017014649.1 960 Silent Mutation CTG,CTT L166L XP_016870138.1
XM_017014650.1 960 Silent Mutation CTG,CTT L33L XP_016870139.1

View Full Product Details