Product Details

SNP ID
rs201823062
Assay Type
Functionally Tested
NCBI dbSNP Submissions
3
Location
Chr.9:104694623 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCCTGCTCTGCCTCATCATGTAC[A/G]TGATAATCCTCCTGGGAAATAGCCT
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
OR13D1 PubMed Links

Gene Details

Gene
OR13D1
Gene Name
olfactory receptor family 13 subfamily D member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004484.1 202 Missense Mutation ATG,GTG M68V NP_001004484.1

View Full Product Details