Product Details

SNP ID
rs200342072
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48576540 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACCAGGGCTATGGGAGTGGCAGG[G/T]ATTATGACAGTCGACCTGGAGGGTA
Phenotype
MIM: 300027
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RBM3 PubMed Links

Gene Details

Gene
RBM3
Gene Name
RNA binding motif (RNP1, RRM) protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006743.4 583 Missense Mutation GAT,TAT D117Y NP_006734.1

View Full Product Details